Lead(II) nitrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:10099-74-8
:
:Pb(NO₃)₂
:MFCD00011153
:331.21 g/mol
:13,05432
:450…470 °C
:4.53 g/cm³ (20 °C)
:Ambient
:233-245-9
:5.1 (6.1),II
:M-Clarity™ quality level = MQ300
:1469
:GHS05
:GHS08
:GHS07
:GHS09
- Solubility:522 g/l (20 °C)
- Melting Point:458 - 459 °C
- Molar Mass:331.2 g/mol
- Bulk Density:1850 kg/m3
- Vapor Pressure:(20 °C)
- Density:4.49 g/cm3 (20 °C)
- pH:3 - 4 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Frequently Bought Together