Order Entry
Czech Republic
ContactUsLinkComponent
Boron nitride, Sigma-Aldrich®
Boron nitride, Sigma-Aldrich®
Catalog # 1.12073.0050
Supplier:  Merck
CAS Number:  
Boron nitride, Sigma-Aldrich®
Catalog # 1.12073.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    2700 - 3000 °C
  • Molar Mass:
    24.82 g/mol
  • Bulk Density:
    120 kg/m3
  • Density:
    2.34 g/cm3 (20 °C)
  • pH:
    5 - 8 (100 g/l, H2O, 20 °C) (slurry)

Specification Test Results

About this item

 :
 :24.82 g/mol
Merck Index:13,01336
Melting Pt:2527 °C
Density:2.15 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:233-136-6
CAS Number:10043-11-5
Formula:BN
MDL Number:MFCD00011317