Boron nitride ≥99.5% (metals basis)
Catalog # 44838.KG
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:24.82 g/mol
Merck Index:14,1346
Melting Pt:2527 °C
Density:2.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-136-6
CAS Number:10043-11-5
Formula:BN
MDL Number:MFCD00011317