Boron nitride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10043-11-5
:
Formula:BN
MDL Number:MFCD00011317
:24.82 g/mol
Merck Index:13,01336
Melting Pt:2527 °C
Density:1.40
EINECS:233-136-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together