Magnesium sulphate, (dried basis) ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7487-88-9
:
Formula:MgSO₄
MDL Number:MFCD00011110
:120.37 g/mol
Merck Index:12,05731
Melting Pt:1127 °C
Density:2.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
Specification Test Results
- Assay:Min. 98 %
- Heavy metals (as Pb):Max. 10 ppm
- Cl (Chloride):Max. 0.05 %
- Fe (Iron):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 33L Plastic bucket UN Z (ZPACK13247037) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Informace o balení
291184P_MAGNESIUM SULPHATE DRIED GPR RECTAPUR 291186R_MAGNESIUM SULPHATE DRIED GPR RECTAPUR_BSE_REV0
Certifikáty BSE/TSE
spec_VWRC291184P spec_VWRC291184P spec_VWRC291186R spec_VWRC291186R spec_VWRC29118BW
Specifikace produktu
více
VWR Chemicals Big Packs and Bulks VWR® for Drying and Filtration
Produktová literatura
Frequently Bought Together