Calcium hydrogen orthophosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7757-93-9
:
Formula:CaHPO₄
MDL Number:MFCD00010909
:136.06 g/mol
Merck Index:13,01697
Melting Pt:370 °C
Density:2.89 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:231-826-1
Frequently Bought Together