Magnesium hydroxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1309-42-8
:
Formula:Mg(OH)₂
MDL Number:MFCD00011104
Density:2.3261…2.3631 g/cm³ (20 °C)
Molecular Weight:58.32 g/mol
Merck Index:13,05693
:M-Clarity™ quality level = MQ200
EINECS:215-170-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together