Magnesium oxide light ≥97%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1309-48-4
:
Formula:MgO
MDL Number:MFCD00011109
:40.3 g/mol
Boiling Pt:3600 °C (1013 hPa)
Melting Pt:2800 °C
Density:3.58 g/cm³ (25 °C)
EINECS:215-171-9
Specification Test Results
- Assay (on calcinated product):Min. 97 %
- Loss on ignition:Max. 8 %
- Silica:Max. 1.5 %
Specification Test Results
Frequently Bought Together