

Barium oxide ≥99.5% (metals basis excluding Sr) (max. 0.05% Sr)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1304-28-5
:12231-51-5
Formula:BaO
MDL Number:MFCD00003453
:153.33 g/mol
Merck Index:14,986
Boiling Pt:∼2000 °C (1013 hPa)
Melting Pt:1923 °C
Density:5.7
Storage Temperature:Ambient
EINECS:215-127-9
UN:1884
ADR:6.1,III
:GHS07
:GHS05
Related Information
Frequently Bought Together