
Barium iodide, anhydrous ≥99.995% (trace metals basis), beads −10 mesh, Sigma-Aldrich®
Dodavatel: Merck
Certifikáty
O tomto produktu
Číslo CAS:13718-50-8
:
Vzorec:BaI₂
MDL číslo:MFCD00003452
:391.14 g/mol
Merck Index:12,01009
Bod tání:714 °C
Hustota:5.15 g/cm³ (20 °C)
EINECS:237-276-9
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:1564
:GHS07
Frequently Bought Together