ContactUsLinkComponent
Calcium carbonate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC (max. 0.002% Fe), precipitated, SAFC®
Calcium carbonate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC (max. 0.002% Fe), precipitated, SAFC®
  1.02064.9029
 :  
Calcium carbonate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC (max. 0.002% Fe), precipitated, SAFC®
  1.02064.9029
 :  
  • Pk:
    25 kg
  • Seq:
    4283
 :
 :100.09 g/mol
 :13,01658
 :825 °C
 :2.71 g/cm³ (20 °C)
 :Ambient
 :207-439-9
 :471-34-1
 :CaCO₃
 :MFCD00010906