Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium hydrogen difluoride ≥98%, GPR RECTAPUR®
Toxic
Corrosive
Ammonium hydrogen difluoride ≥98%, GPR RECTAPUR®
Catalog # 21262.360
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Corrosive
Ammonium hydrogen difluoride ≥98%, GPR RECTAPUR®
Catalog # 21262.360
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 0.03 %
  • Cl (Chloride):
    Max. 0.05 %
  • SiF6 (Hexafluorosilicate):
    Max. 0.3 %
  • Fe (Iron):
    Max. 100 ppm

Specification Test Results

About this item

 :
 :57.04 g/mol
Merck Index:13,00498
Melting Pt:127 °C
Density:0.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-676-4
UN:1727
ADR:8,II
 :GHS06
 :GHS05
CAS Number:1341-49-7
Formula:NH₄HF₂
MDL Number:MFCD00012018