

Potassium hydrogen sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
0198.0500
: Avantor
:
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Acidity (as H2SO4):35.0 - 37.0 %
- Calcium and Magnesium Precipitate:max. 0.005 %
- Chloride (Cl):max. 0.001 %
- Iron (Fe):max. 0.001 %
- Heavy Metals (as Pb):max. 0.001 %
- Insoluble Matter and NH4OH Precipitate:max. 0.01 %
- Phosphate (PO4):max. 0.001 %
- Arsenic (As):max. 1
:
:13,07695
:214 °C
:2.32 g/cm³ (20 °C)
:8,II
:136.17 g/mol
:231-594-1
:2509
:GHS05
:GHS07
:7646-93-7
:KHSO₄
:MFCD00011404