

Disodium metasilicate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6834-92-0
:
Formula:Na₂O₃Si
MDL Number:MFCD00003492
:122.06 g/mol
Merck Index:12,08788
Melting Pt:∼72.2 °C
Density:1.749 g/cm³ (20 °C)
EINECS:229-912-9
ADR:8,III
:M-Clarity™ quality level = MQ200
UN:3253
:GHS05
:GHS07
Frequently Bought Together