
Potassium thiocyanate 99+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
Merck Index:12,07866
Boiling Pt:500 °C (1013 hPa)
Melting Pt:170…179 °C
Density:1.886 g/cm³
:97.18 g/mol
EINECS:206-370-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Titration Argentometric:≥99.0 %
- pH:5.3 to 8.7 (5 % at 20°C)
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Ammonium (NH4):≤10 ppm
- Iron (Fe):≤2 ppm
- Sodium (Na):≤200 ppm
- Heavy metals (as Pb):≤2 ppm
- Iodine consuming subst.:≤30 mg I2/100 g
Specification Test Results
Frequently Bought Together