Potassium dihydrogen phosphate 98.0-100.5%, crystallised Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Merck Index:13,07744
Boiling Pt:~100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
Specification Test Results
- Assay (calculated on dried substance):98.0 - 100.5 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification B:Passes test
- Identification C:Passes test
- Appearance of solution:Passes test
- pH (5 %):4.2 - 4.5
- Reducing substances:Passes test
- Cl (Chloride):Max. 200 ppm
- SO4 (Sulphate):Max. 300 ppm
- As (Arsenic):Max. 2 ppm
- Fe (Iron):Max. 10 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Loss on drying (125-130°C):Max. 2.0 %
- Residual solvents:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 30L Plastic bucket UN Z (ZPACK13247037) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
26922_POTASSIUM DIHYDROGEN PHOSPHATE CRYS PH.EUR._Latex_REV0.pdf 26922_POTASSIUM DIHYDROGEN PHOSPHATE CRYS PH.EUR._Melamine_REV0.pdf 26922_POTASSIUM DIHYDROGEN PHOSPHATE CRYS PH.EUR._Nanomaterials_REV0.pdf 26922_POTASSIUM DIHYDROGEN PHOSPHATE CRYS PH.EUR._Phtalate_REV0.pdf Potassium Dihydrogen Phosphate 98 - 100,5%, Crystallised Ph. Eur.
Frequently Bought Together