

Orthophosphoric acid ≥85-88% NF, ACS, J.T.Baker®
0240-07
: Avantor
:
- Pk:700 lb
- Seq:7365
- Bulk Pharmaceutical Chemical:85.0 - 88.0 %
- NF - Assay (H₃PO₄):85.0 - 88.0 %
- NF - Identification:Passes Test
- NF - Nitrate (NO₃):Passes Test
- NF - Phosphorous or Hypophosphorous Acid:Passes Test
- NF - Sulfate (SO₄):Passes Test
- NF - Alkali Phosphates:Passes Test
- Assay (H₃PO₄) (by acidimetry):85.0 - 87.0 %
- Calcium (Ca):≤0.002 %
- Chloride (Cl):≤3 ppm
- Color (APHA):≤10
- Heavy Metals (as Pb):≤8 ppm
- Insoluble Matter:≤0.001 %
- ACS - Magnesium (Mg):≤0.002 %
- Nitrate (NO₃):≤5 ppm
- Reducing Substances:Passes Test
- Sulfate (SO₄):≤12 ppm
- Trace Impurities - Antimony (Sb):≤20.000 ppm
- Trace Impurities - Arsenic (As):≤0.500 ppm
- Trace Impurities - Iron (Fe):≤10 ppm
- Trace Impurities - Manganese (Mn):≤0.500 ppm
- Trace Impurities - Potassium (K):≤40.000 ppm
- Trace Impurities - Sodium (Na):≤200.000 ppm
- Volatile Acids (as CH₃COOH):≤0.001 %
:
:98 g/mol
:13,07744
:~ 100 °C (1013 hPa)
:253 °C
:2.3 g/cm³ (20 °C)
:Ambient
:231-913-4
:1805
:8,III
:01-2119490224-41
:GHS05
:GHS07
:7664-38-2
:H₃PO₄
:MFCD00011340