
di-Potassium hydrogen phosphate ≥98.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-11-4
:
Formula:K₂HPO₄
MDL Number:MFCD00011383
:174.18 g/mol
Merck Index:14,7658
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
EINECS:231-834-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together