Sodium metaphosphate, GPR RECTAPUR®
Plastic bottle for solids, 1 kg
Supplier: VWR Chemicals
Sodium metaphosphate, GPR RECTAPUR®
Catalog # 27930.290
Supplier: VWR Chemicals
CAS Number: 10361-03-2
Jump to:
Product Details & Documents
- CAS Number:10361-03-2
- Formula:NaPO₃
- Molecular Weight:101.96 g/mol
- Melting Pt:625 °C
- Density:2.54 g/cm³ (20 °C)
- :Density: 2.54 g/cm³ (20 °C)
- EINECS:233-782-9
Specifications
Specification Test Results
- di-Phosphorus pentoxide:Min. 60 %
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 0.05 %
- Fe (Iron):Max. 50 ppm
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg



