Order Entry
Czech Republic
ContactUsLinkComponent
penta-Sodium triphosphate for BOD determination acc. to DIN 38409-H-51, Supelco®
penta-Sodium triphosphate for BOD determination acc. to DIN 38409-H-51, Supelco®
Catalog # 1.06999.1000
Supplier:  Merck
CAS Number:  
penta-Sodium triphosphate for BOD determination acc. to DIN 38409-H-51, Supelco®
Catalog # 1.06999.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (acidimetric, P₂O₅):
    57 - 59 %
  • pH-value (1 %; water):
    43382
  • Fe (Iron):
    ≤ 0.005 %

Specification Test Results

About this item

 :
 :367.86 g/mol
Merck Index:12,08846
Melting Pt:622 °C (dec.)
Density:2.52 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-838-7
CAS Number:7758-29-4
Formula:Na₅O₁₀P₃
MDL Number:MFCD00003514