
Potassium nitrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7757-79-1
:
Formula:KNO₃
MDL Number:MFCD00011409
:101.1 g/mol
Merck Index:13,07733
Melting Pt:334 °C
Density:2.109 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-818-8
ADR:5.1,III
:M-Clarity™ quality level = MQ200
UN:1486
REACH:01-2119488224-35
:GHS03
Frequently Bought Together