Lithium niobate, Sigma-Aldrich®
Catalog # SIAL254290-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:147.85 g/mol
Melting Pt:1257 °C
Density:4.638 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:234-755-4
CAS Number:12031-63-9
Formula:LiNbO₃
MDL Number:MFCD00011080