Lithium iodide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10377-51-2
:
Formula:ILi
MDL Number:MFCD00011092
:133.85 g/mol
Merck Index:13,05557
Boiling Pt:1177 °C (1013 hPa)
Melting Pt:450 °C
Density:3.49 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:233-822-5
Frequently Bought Together