Order Entry
Czech Republic
ContactUsLinkComponent
Cesium iodide, Sigma-Aldrich®
Irritant
Cesium iodide, Sigma-Aldrich®
Catalog # SIAL429384-10G
Supplier:  Merck
CAS Number:  
Irritant
Cesium iodide, Sigma-Aldrich®
Catalog # SIAL429384-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :259.81 g/mol
Merck Index:12,02056
Boiling Pt:1280 °C (1013 hPa)
Melting Pt:621 °C
Density:3.21 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:232-145-2
 :GHS07
CAS Number:7789-17-5
Formula:CsI
MDL Number:MFCD00010962