
Cesium iodide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-17-5
:
Formula:CsI
MDL Number:MFCD00010962
:259.81 g/mol
Merck Index:14,2014
Boiling Pt:1280 °C
Melting Pt:626 °C
Density:3.21 g/cm³ (20 °C)
EINECS:232-145-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together