



Sodium cyanotrihydroborate ≥95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25895-60-7
:
Formula:CH₃BNNa
MDL Number:MFCD00003516
:62.84 g/mol
Merck Index:14,8606
Melting Pt:>300 °C (dec.)
Density:1.20
EINECS:247-317-2
UN:3134
ADR:4.3,II
:GHS02
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together