Order Entry
Czech Republic
ContactUsLinkComponent
Potassium hexacyanoferrate(III) ≥96%, TECHNICAL
Potassium hexacyanoferrate(III) ≥96%, TECHNICAL
Catalog # 26807.297
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium hexacyanoferrate(III) ≥96%, TECHNICAL
Catalog # 26807.297
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 96 %

Specification Test Results

About this item

 :
 :329.25 g/mol
Merck Index:13,07712
Density:1.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-323-3
CAS Number:13746-66-2
Formula:K₃Fe(CN)₆
MDL Number:MFCD00011392