

Cesium fluoride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13400-13-0
:
Formula:CsF
MDL Number:MFCD00010960
:151.9 g/mol
Boiling Pt:1251 °C (1013 hPa)
Melting Pt:680 °C
Density:4.627 g/cm³ (20 °C)
EINECS:236-487-3
UN:2923
ADR:8,III
:GHS06
:GHS05
Frequently Bought Together