

Cesium fluoride ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13400-13-0
:
Formula:CsF
MDL Number:MFCD00010960
:151.9 g/mol
Boiling Pt:1251 °C
Melting Pt:682 °C
Density:4.627 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-487-3
UN:2923
ADR:8,II
:GHS06
:GHS05
Related Information
Frequently Bought Together