Order Entry
Czech Republic
ContactUsLinkComponent
Cesium fluoride 99% for analysis
Corrosive
Toxic
Health
Cesium fluoride 99% for analysis
Certificates
undefined
Corrosive
Toxic
Health
Cesium fluoride 99% for analysis

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Assay:
    ≥98.5 %
  • Chloride (Cl):
    ≤200 ppm
  • Sulfate (SO4):
    ≤300 ppm
  • Aluminium (Al):
    ≤10 ppm
  • Barium (Ba):
    ≤10 ppm
  • Calcium (Ca):
    ≤50 ppm
  • Lithium (Li):
    ≤5 ppm
  • Magnesium (Mg):
    ≤50 ppm
  • Potassium (K):
    ≤300 ppm
  • Rubidium (Rb):
    ≤200 ppm
  • Sodium (Na):
    ≤100 ppm
  • Strontium (Sr):
    ≤10 ppm

Specification Test Results

Related Information

Frequently Bought Together