Lithium bromide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7550-35-8
:23303-71-1
Formula:LiBr
MDL Number:MFCD00011077
Boiling Pt:1265
Melting Pt:550
Density:3.46 g/cm³ (20 °C)
Molecular Weight:86.84 g/mol
Merck Index:13,05548
:M-Clarity™ quality level = MQ100
EINECS:231-439-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together