Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cesium bromide
Cesium bromide ≥99% (metals basis)
Cesium bromide ≥99% (metals basis)
:
Thermo Fisher Scientific
Cesium bromide ≥99% (metals basis)
:
Thermo Fisher Scientific
:
7787-69-1
:
:
BrCs
:
MFCD00010954
:
212.81 g/mol
:
14,2009
:
1300 °C
:
636 °C
:
4.44
:
232-130-0
:
GHS07
:
Ambient
msds_ALFA013490
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Methanol, HiPerSolv CHROMANORM®, Ultra Plus for LC-MS, suitable for UPLC/UHPLC-MS instruments
: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone, EMPARTA® ACS analytical reagent, Supelco®
: Merck
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Tetrahydrofuran, Uvasol® for spectroscopy, Supelco®
: Merck
Water, GPR RECTAPUR®
: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Methyl ethyl ketone, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals