Order Entry
Czech Republic
ContactUsLinkComponent
Lithium metaborate, Sigma-Aldrich®
Lithium metaborate, Sigma-Aldrich®
Catalog # SIAL254282-10G
Supplier:  Merck
CAS Number:  
Lithium metaborate, Sigma-Aldrich®
Catalog # SIAL254282-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :49.75 g/mol
Merck Index:13,05546
Boiling Pt:1719 °C (1013 hPa)
Melting Pt:845 °C
Density:1.4 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:236-631-5
CAS Number:13453-69-5
Formula:BLiO₂
MDL Number:MFCD00011089