Order Entry
Czech Republic
ContactUsLinkComponent
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Colouration:
    Max. 5 APHA
  • Conductivity (25°C) (at filling):
    Max. 1 µS/cm
  • Evaporation residue:
    Max. 1 ppm
  • Al (Aluminium):
    Max. 50 ppb
  • Ca (Calcium):
    Max. 50 ppb
  • Fe (Iron):
    Max. 50 ppb
  • K (Potassium):
    Max. 50 ppb
  • Mg (Magnesium):
    Max. 50 ppb
  • Na (Sodium):
    Max. 100 ppb
  • Fluorescence (as quinine) (254/450 nm):
    Max. 1 ppb
  • Fluorescence (as quinine) (365/450 nm):
    Max. 0.5 ppb
  • Gradient grade (210 nm):
    Max. 5 mAU
  • Gradient (220 nm):
    Max. 2 mAU
  • Gradient (254 nm):
    Max. 1 mAU
  • MS-ESI+ (as Reserpine):
    Max. 2 ppb
  • MS-APCI+ (as Reserpine):
    Max. 2 ppb
  • MS-ESI- (as 4-Nitrophenol):
    Max. 20 ppb
  • MS-APCI- (as 4-Nitrophenol):
    Max. 20 ppb
  • Colony count (Ph.Eur.) (at filling):
    Max. 100 CFU/ml
  • TOC (at filling):
    Max. 30 ppb

Specification Test Results

Frequently Bought Together