1,4-Dihydroxy-9,10-anthraquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:81-64-1
:
Formula:C₁₄H₈O₄
MDL Number:MFCD00001209
:240.22 g/mol
Merck Index:12,08250
Boiling Pt:465 °C (1013 hPa)
Melting Pt:202 °C
Density:1.278 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:201-368-7
Specification Test Results
- Solubility:- 1 g/l (20 °C)
- Melting Point:195 - 198 °C
- Molar Mass:240.21 g/mol
- Bulk Density:350 kg/m3
- Boiling Point:450 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (197 °C)
- Flash Point:222 °C
Specification Test Results
Frequently Bought Together