2,7-Dibromo-9-fluorenone, Sigma-Aldrich®
Catalog # SIAL300098-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:338 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:14348-75-5
Formula:C₁₃H₆Br₂O
MDL Number:MFCD00010790