

4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222), Kryptofix® for synthesis, Sigma-Aldrich®
Catalog # 8.14925.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Melting Point:68 - 71 °C
- Molar Mass:376.49 g/mol
Specification Test Results
About this item
:
:376.49 g/mol
Melting Pt:69…71 °C
:M-Clarity™ quality level = MQ200
EINECS:245-962-4
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:23978-09-8
Formula:C₁₈H₃₆N₂O₆
MDL Number:MFCD00005111