

2-Cyclopenten-1-one ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:930-30-3
:
Formula:C₅H₆O
MDL Number:MFCD00001401
:82.1 g/mol
Boiling Pt:150 °C
Density:1.01
Flash Pt:42 °C (108 °F)
Storage Temperature:Refrigerator
EINECS:213-213-0
UN:1224
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together