

2-Cyclopenten-1-one 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:930-30-3
:
Formula:C₅H₆O
MDL Number:MFCD00001401
:82.1 g/mol
Boiling Pt:150 °C (760 mmHg)
Density:0.98 g/cm³
Flash Pt:42 °C
Storage Temperature:Refrigerator
EINECS:213-213-0
UN:1224
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4800 to 1.4830 (20°C, 589 nm)
- GC:≥97.5 %
- UV:(2.7 g in 50 ml water) Acidified to pH 3 with Phosphoric Acid ACS grade
Specification Test Results
Frequently Bought Together