Přeskočit na Záhlaví
Přeskočit na Hlavní obsah
Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Cyclohexen-1-one
2-Cyclohexen-1-one ≥97%
2-Cyclohexen-1-one ≥97%
:
Thermo Fisher Scientific
2-Cyclohexen-1-one ≥97%
:
Thermo Fisher Scientific
:
930-68-7
:
:
C₆H₈O
:
MFCD00001577
:
96.13 g/mol
:
167…169 °C
:
–53 °C
:
0.993
:
56 °C (132 °F)
:
Ambient
:
213-223-5
:
2929
:
6.1,II
:
GHS06
:
GHS02
:
GHS07
msds_ALFAA15435
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
n-Hexane, dehydrated (max. 0.01% H₂O) ≥95%, GPR RECTAPUR® for synthesis
: VWR Chemicals
2-Propanol, LiChrosolv®, gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitrile ≥99.95%, HiPerSolv CHROMANORM® ULTRA for LC-MS, suitable for UPLC/UHPLC-MS instruments
: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Dichloromethane stabilised Ph. Eur.
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
Hanus solution (Iodine bromine) c(IBr) = 0.1 mol/l, Titripur® for determination of iodine value, Supelco®
: Merck
Acrylonitrile 99+%
: Thermo Fisher Scientific