Tetramethyl-1,3-cyclobutanedione, Sigma-Aldrich®
Catalog # SIALT21008-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:140.18 g/mol
:M-Clarity™ quality level = MQ100
EINECS:213-269-6
CAS Number:933-52-8
Formula:C₈H₁₂O₂
MDL Number:MFCD00001331