
5-Methyl-1,3-cyclohexanedione, Sigma-Aldrich®
Catalog # SIAL279951-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:126.16 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
Storage Temperature:Ambient
CAS Number:4341-24-6
Formula:C₇H₁₀O₂
MDL Number:MFCD00010379