2-Methyl-1,3-cyclohexanedione, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1193-55-1
:
Formula:C₇H₁₀O₂
MDL Number:MFCD00001587
Boiling Pt:100…102 °C (8 mmHg)
Melting Pt:206…209 °C
Density:1.313 g/cm³ (–150.15 °C)
:M-Clarity™ quality level = MQ100
:126.16 g/mol
Storage Temperature:Ambient
EINECS:214-773-9
Frequently Bought Together