

2-Methylcyclohexanone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:583-60-8
:
Formula:C₇H₁₂O
MDL Number:MFCD00001635
:112.17 g/mol
Boiling Pt:163 °C (1013 hPa)
Melting Pt:–14 °C
Density:0.924 g/cm³ (20 °C)
Flash Pt:48 °C
UN:2297
ADR:3,III
:M-Clarity™ quality level = MQ100
EINECS:209-513-6
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together