Order Entry
Czech Republic
ContactUsLinkComponent
 
Cyclohexyl isothiocyanate, Sigma-Aldrich®
Catalog # SIALC105406-25G
Supplier:  Merck
CAS Number:  
Cyclohexyl isothiocyanate, Sigma-Aldrich®
Catalog # SIALC105406-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :141.24 g/mol
Boiling Pt:219 °C (1013 hPa)
Melting Pt:70…72 °C
Density:1.0339 g/cm³ (20 °C)
EINECS:214-361-9
UN:3265
ADR:8,II
 :M-Clarity™ quality level = MQ100
CAS Number:1122-82-3
Formula:C₇H₁₁NS
MDL Number:MFCD00003841