1,3-Cyclohexanedione, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:504-02-9
:
Formula:C₆H₈O₂
MDL Number:MFCD00001585
:112.13 g/mol
Merck Index:13,03200
Boiling Pt:235 °C (1013 hPa)
Melting Pt:105 °C
Density:1.25 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:207-980-0
Frequently Bought Together