Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Cyclohexanone, Sigma-Aldrich®

Flammable
Irritant
   Sustainable Options Available
Supplier:   Merck
Cyclohexanone, Sigma-Aldrich®
Supplier:  Merck
   Sustainable Options Available
Certificates
Cyclohexanone, Sigma-Aldrich®

4 Options Available Below

Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35

 

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SIALC102180-25ML
  • SIALC102180-1L
  • SIALC102180-2.5L
  • SIALC102180-25L
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 963,00 Kč
    963,00 Kč
    1 * 25 mL
       963,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SIALC102180-25ML
    Environmentally Preferable
    Environmental certification
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 1 730,00 Kč
    1 730,00 Kč
    1 * 1 L
       1 730,00 Kč

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SIALC102180-1L
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 4 290,00 Kč
    4 290,00 Kč
    1 * 2,5 L
       4 290,00 Kč

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SIALC102180-2.5L
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
  • Glass bottle
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 35 630,00 Kč
    35 630,00 Kč
    1 * 25 L
       35 630,00 Kč

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SIALC102180-25L
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health