Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Cyclohexanone, Sigma-Aldrich®

Cyclohexanone, Sigma-Aldrich®

Flammable
Irritant
Supplier:   Merck
Cyclohexanone, Sigma-Aldrich®
Supplier:  Merck
Certificates
Cyclohexanone, Sigma-Aldrich®
Cyclohexanone, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 745,00 Kč
    745,00 Kč
    1 * 25 mL
       745,00 Kč

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1 480,00 Kč
    1 480,00 Kč
    1 * 500 mL
       1 480,00 Kč

  • Glass bottle
    2 L
    Log in for availability
    Log in for availability
    1 * 2 L
    1 * 2 L - 4 890,00 Kč
    4 890,00 Kč
    1 * 2 L
       4 890,00 Kč

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.