cis,cis-1,3-Cyclooctadiene, Sigma-Aldrich®
Catalog # SIAL274100-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:108.18 g/mol
Boiling Pt:143 °C (1013 hPa)
Melting Pt:–53…–51 °C
Density:0.869 g/cm³ (20 °C)
Storage Temperature:Refrigerator
UN:2520
ADR:3,III
:M-Clarity™ quality level = MQ100
CAS Number:3806-59-5
Formula:C₈H₁₂
MDL Number:MFCD00001751