
2-Methyl-8-quinolinol, Sigma-Aldrich®
Catalog # SIALH57602-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:159.19 g/mol
Boiling Pt:267 °C (1013 hPa)
Melting Pt:70…72 °C
Storage Temperature:Ambient
ADR:9,III
:M-Clarity™ quality level = MQ100
EINECS:212-562-6
UN:3077
:GHS09
CAS Number:826-81-3
Formula:C₁₀H₉NO
MDL Number:MFCD00006765